Assay Details
Gene Symbol: CYB5RL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001492765.5 | XP_001492815.2 | 880 | 66 | ||
XM_005607026.3 | XP_005607083.1 | 3961 | 66 | |||
XM_005607027.3 | XP_005607084.1 | 882 | 66 | |||
XM_005607028.3 | XP_005607085.1 | 884 | 66 | |||
XM_005607029.3 | XP_005607086.1 | 880 | 66 | |||
XM_014737462.2 | XP_014592948.1 | 891 | 66 | |||
XM_014737463.2 | XP_014592949.1 | 882 | 66 | |||
XM_023629381.1 | XP_023485149.1 | 1148 | 66 | |||
XM_023629391.1 | XP_023485159.1 | 3772 | 66 | |||
XM_023629397.1 | XP_023485165.1 | 492 | 66 | |||
XM_023629406.1 | XP_023485174.1 | 881 | 66 | |||
XM_023629411.1 | XP_023485179.1 | 359 | 66 | |||
XM_023629454.1 | XP_023485222.1 | 882 | 66 | |||
XM_023629477.1 | XP_023485245.1 | 884 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
