Assay Details
Gene Symbol: CPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489650.6 | XP_001489700.1 | 1805 | 54 | ||
XM_014737304.2 | XP_014592790.1 | 1908 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
