Assay Details
Gene Symbol: ZYG11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014737412.2 | XP_014592898.1 | 1328 | 66 | ||
XM_023629944.1 | XP_023485712.1 | 1539 | 66 | |||
XM_023629949.1 | XP_023485717.1 | 1625 | 66 | |||
XM_023629960.1 | XP_023485728.1 | 1366 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
