Assay Details
Gene Symbol: RAB3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023629977.1 | XP_023485745.1 | 478 | 64 | ||
XM_023629982.1 | XP_023485750.1 | 520 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
