Assay Details
Gene Symbol: BEND5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023636831.1 | XP_023492599.1 | 1197 | 87 | ||
XM_023636832.1 | XP_023492600.1 | 1061 | 87 | |||
XM_023636833.1 | XP_023492601.1 | 961 | 87 | |||
XM_023636835.1 | XP_023492603.1 | 958 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
