Assay Details
Gene Symbol: STIL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493358.4 | XP_001493408.2 | 677 | 87 | ||
XM_005607072.3 | XP_005607129.2 | 677 | 87 | |||
XM_014737533.2 | XP_014593019.2 | 677 | 87 | |||
XM_023630338.1 | XP_023486106.1 | 642 | 87 | |||
XM_023630357.1 | XP_023486125.1 | 822 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
