Assay Details
Gene Symbol: TOE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496197.4 | XP_001496247.1 | 125 | 82 | ||
XM_005607104.3 | XP_005607161.1 | 125 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
