Assay Details
Gene Symbol: B4GALT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023631545.1 | XP_023487313.1 | 989 | 60 | ||
XM_023631550.1 | XP_023487318.1 | 950 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
