Assay Details
Gene Symbol: MED8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498503.5 | XP_001498553.2 | 356 | 59 | ||
XM_005607127.3 | XP_005607184.1 | 350 | 59 | |||
XM_014737367.2 | XP_014592853.1 | 350 | 59 | |||
XM_014737369.2 | XP_014592855.1 | 350 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
