Assay Details
Gene Symbol: GUCA2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503167.3 | XP_001503217.1 | 527 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
