Assay Details
Gene Symbol: AK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503750.5 | XP_001503800.1 | 164 | 75 | ||
XM_005607238.3 | XP_005607295.1 | 164 | 75 | |||
XM_023634099.1 | XP_023489867.1 | 182 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
