Assay Details
Gene Symbol: S100PBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005607244.3 | XP_005607301.1 | 1392 | 83 | ||
XM_014737295.2 | XP_014592781.1 | 1205 | 83 | |||
XM_023634197.1 | XP_023489965.1 | 1222 | 83 | |||
XM_023634210.1 | XP_023489978.1 | 1528 | 83 | |||
XM_023634214.1 | XP_023489982.1 | 1218 | 83 | |||
GenBank mRNA | JL625558.1 | - | 3 - 4 | 447 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
