Assay Details
Gene Symbol: ZCCHC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503910.5 | XP_001503960.1 | 595 | 70 | ||
XM_005607260.3 | XP_005607317.1 | 595 | 70 | |||
XM_023634634.1 | XP_023490402.1 | 656 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
