Assay Details
Gene Symbol: YTHDF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500333.5 | XP_001500383.2 | 289 | 60 | ||
XM_014737386.2 | XP_014592872.1 | 322 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
