Assay Details
Gene Symbol: SMPDL3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005615183.3 | XP_005615240.2 | 544 | 66 | ||
XM_023635104.1 | XP_023490872.1 | 541 | 66 | |||
XM_023635109.1 | XP_023490877.1 | 542 | 66 | |||
XM_023635116.1 | XP_023490884.1 | 540 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
