Assay Details
Gene Symbol: CRYBG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014737445.2 | XP_014592931.2 | 4216 | 56 | ||
XM_023635414.1 | XP_023491182.1 | 4216 | 56 | |||
XM_023635415.1 | XP_023491183.1 | 4216 | 56 | |||
XM_023635416.1 | XP_023491184.1 | 4216 | 56 | |||
XM_023635417.1 | XP_023491185.1 | 4216 | 56 | |||
XM_023635418.1 | XP_023491186.1 | 4216 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
