Assay Details
Gene Symbol: MTFR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023635468.1 | XP_023491236.1 | 331 | 72 | ||
XM_023635469.1 | XP_023491237.1 | 316 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
