Assay Details
Gene Symbol: NCMAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504170.6 | XP_001504220.4 | 325 | 117 | ||
XM_005607345.3 | XP_005607402.2 | 437 | 117 | |||
XM_023635491.1 | XP_023491259.1 | 250 | 117 | |||
XM_023635492.1 | XP_023491260.1 | 451 | 117 | |||
XM_023635493.1 | XP_023491261.1 | 337 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
