Assay Details
Gene Symbol: RCAN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005607346.3 | XP_005607403.1 | 740 | 62 | ||
XM_005607347.3 | XP_005607404.1 | 778 | 62 | |||
XM_014737384.2 | XP_014592870.1 | 718 | 62 | |||
XM_023635494.1 | XP_023491262.1 | 770 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
