Assay Details
Gene Symbol: HNRNPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005607364.3 | XP_005607421.1 | 690 | 63 | ||
XM_023635523.1 | XP_023491291.1 | 826 | 63 | |||
XM_023635524.1 | XP_023491292.1 | 829 | 63 | |||
XM_023635525.1 | XP_023491293.1 | 659 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
