Assay Details
Gene Symbol: WNT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023635550.1 | XP_023491318.1 | 519 | 91 | ||
XM_023635551.1 | XP_023491319.1 | 479 | 91 | |||
XM_023635552.1 | XP_023491320.1 | 2001 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
