Assay Details
Gene Symbol: MUL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504339.4 | XP_001504389.1 | 467 | 69 | ||
XM_005607391.3 | XP_005607448.1 | 669 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
