Assay Details
Gene Symbol: PLA2G2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501688.3 | XP_001501738.2 | 147 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001501688.3 transcripts.
Set Membership: |
![]() ![]() |
