Assay Details
Gene Symbol: GPR162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001915670.4 | XP_001915705.1 | 1781 | 87 | ||
XM_005610886.3 | XP_005610943.1 | 1781 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
