Assay Details
Gene Symbol: ATN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023643062.1 | XP_023498830.1 | 483 | 91 | ||
XM_023643063.1 | XP_023498831.1 | 483 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
