Assay Details
Gene Symbol: SLC2A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498757.4 | XP_001498807.1 | 1113 | 66 | ||
XM_023643089.1 | XP_023498857.1 | 998 | 66 | |||
XM_023643090.1 | XP_023498858.1 | 1206 | 66 | |||
XM_023643091.1 | XP_023498859.1 | 1086 | 66 | |||
XM_023643092.1 | XP_023498860.1 | 1043 | 66 | |||
GenBank mRNA | JL632303.1 | - | 2 - 3 | 129 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
