Assay Details
Gene Symbol: CLEC7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005610931.3 | XP_005610988.2 | 404 | 79 | ||
XM_005610932.3 | XP_005610989.2 | 406 | 79 | |||
XM_014740671.2 | XP_014596157.2 | 262 | 79 | |||
XM_014740672.2 | XP_014596158.1 | 253 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
