Assay Details
Gene Symbol: DUSP16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023643138.1 | XP_023498906.1 | 1461 | 76 | ||
XM_023643139.1 | XP_023498907.1 | 1591 | 76 | |||
XM_023643140.1 | XP_023498908.1 | 1023 | 76 | |||
XM_023643141.1 | XP_023498909.1 | 1000 | 76 | |||
XM_023643142.1 | XP_023498910.1 | 5115 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
