Assay Details
Gene Symbol: CREBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501394.5 | XP_001501444.1 | 539 | 68 | ||
XM_014740665.2 | XP_014596151.1 | 597 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
