Assay Details
Gene Symbol: GPRC5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501469.3 | XP_001501519.1 | 1091 | 68 | ||
XM_005610949.3 | XP_005611006.1 | 1348 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
