Assay Details
Gene Symbol: GOLT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023643220.1 | XP_023498988.1 | 405 | 90 | ||
GenBank mRNA | JL616452.1 | - | 3 - 4 | 398 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
