Assay Details
Gene Symbol: FBLIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023635709.1 | XP_023491477.1 | 1010 | 63 | ||
XM_023635710.1 | XP_023491478.1 | 1010 | 63 | |||
XM_023635712.1 | XP_023491480.1 | 1109 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
