Assay Details
Gene Symbol: DISP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001915116.4 | XP_001915151.2 | 4111 | 83 | ||
XM_023635798.1 | XP_023491566.1 | 4416 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
