Assay Details
Gene Symbol: DFFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001490534.4 | XP_001490584.1 | 688 | 58 | ||
XM_023635813.1 | XP_023491581.1 | 694 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
