Assay Details
Gene Symbol: CTNNBIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023635829.1 | XP_023491597.1 | 430 | 74 | ||
XM_023635830.1 | XP_023491598.1 | 351 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
