Assay Details
Gene Symbol: PARK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495398.5 | XP_001495448.1 | 511 | 92 | ||
XM_005607554.2 | XP_005607611.1 | 334 | 92 | |||
XM_023635859.1 | XP_023491627.1 | 530 | 92 | |||
XM_023635860.1 | XP_023491628.1 | 336 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
