Assay Details
Gene Symbol: GPR153 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023635888.1 | XP_023491656.1 | 439 | 57 | ||
XM_023635889.1 | XP_023491657.1 | 619 | 57 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023635888.1, XM_023635889.1 transcripts.
Set Membership: |
![]() ![]() |
