Assay Details
Gene Symbol: GPR153 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023635887.1 | XP_023491655.1 | 1407 | 105 | ||
XM_023635888.1 | XP_023491656.1 | 1533 | 105 | |||
XM_023635889.1 | XP_023491657.1 | 1713 | 105 | |||
XM_023635890.1 | XP_023491658.1 | 1557 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
