Assay Details
Gene Symbol: PHYHIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491346.4 | XP_001491396.1 | 1000 | 66 | ||
XM_005607664.3 | XP_005607721.1 | 739 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
