Assay Details
Gene Symbol: TNFRSF10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005607681.3 | XP_005607738.2 | 474 | 63 | ||
XM_014737745.2 | XP_014593231.2 | 473 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
