Assay Details
Gene Symbol: LOXL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001914854.5 | XP_001914889.3 | 1732 | 59 | ||
XM_023636161.1 | XP_023491929.1 | 1755 | 59 | |||
XM_023636162.1 | XP_023491930.1 | 1847 | 59 | |||
XM_023636163.1 | XP_023491931.1 | 1752 | 59 | |||
XM_023636164.1 | XP_023491932.1 | 1757 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
