Assay Details
Gene Symbol: ADAMDEC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005607688.3 | XP_005607745.1 | 984 | 65 | ||
XM_014737821.2 | XP_014593307.1 | 983 | 65 | |||
XM_023636174.1 | XP_023491942.1 | 982 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
