Assay Details
Gene Symbol: BNIP3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494222.5 | XP_001494272.1 | 452 | 89 | ||
XM_005607693.3 | XP_005607750.1 | 454 | 89 | |||
XM_014737787.2 | XP_014593273.1 | 345 | 89 | |||
XM_023636189.1 | XP_023491957.1 | 279 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
