Assay Details
Gene Symbol: ADRA1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014737800.2 | XP_014593286.1 | 2414 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
