Assay Details
Gene Symbol: SCARA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023636196.1 | XP_023491964.1 | 313 | 89 | ||
XM_023636197.1 | XP_023491965.1 | 245 | 89 | |||
XM_023636198.1 | XP_023491966.1 | 314 | 89 |
