Assay Details
Gene Symbol: SCARA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023636196.1 | XP_023491964.1 | 534 | 85 | ||
XM_023636197.1 | XP_023491965.1 | 466 | 85 | |||
XM_023636198.1 | XP_023491966.1 | 535 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
