Assay Details
Gene Symbol: PBK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001492945.4 | XP_001492995.1 | 787 | 72 | ||
GenBank mRNA | JL620876.1 | - | 1 - 2 | 71 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
