Assay Details
Gene Symbol: ZNF395 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023636209.1 | XP_023491977.1 | 1019 | 59 | ||
XM_023636210.1 | XP_023491978.1 | 909 | 59 | |||
XM_023636211.1 | XP_023491979.1 | 947 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
