Assay Details
Gene Symbol: FZD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023636219.1 | XP_023491987.1 | 663 | 65 | ||
XM_023636220.1 | XP_023491988.1 | 658 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
