Assay Details
Gene Symbol: MSRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014737807.2 | XP_014593293.2 | 427 | 93 | ||
XM_023636239.1 | XP_023492007.1 | 553 | 93 | |||
XM_023636240.1 | XP_023492008.1 | 849 | 93 | |||
XM_023636241.1 | XP_023492009.1 | 848 | 93 | |||
XM_023636242.1 | XP_023492010.1 | 845 | 93 | |||
XM_023636243.1 | XP_023492011.1 | 845 | 93 | |||
XM_023636244.1 | XP_023492012.1 | 440 | 93 | |||
XM_023636245.1 | XP_023492013.1 | 785 | 93 | |||
XM_023636246.1 | XP_023492014.1 | 844 | 93 | |||
GenBank mRNA | JL626684.1 | - | 1 - 2 | 106 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
